Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAQRKNAKSS GNSSSSGSGS GSTSAGSSSP GARRETKHGG HKNGRKGGLS GTSFFTWFMV IALLGVWTSV AVVWFDLVDY EEVLGKLGIY DADGDGDFDV DDAKVLLGLK ERSTSEPAVP PEEAEPHTEP EEQVPVEAEP QNIEDEAKEQ IQSLLHEMVH AEHETEHSYH VEETVSQDCN QDMEEMMSEQ ENPDSSEPVV EDERLHHDTD DVTYQVYEEQ AVYEPLENEG IEITEVTAPP EDNPVEDSQV IVEEVSIFPV EEQQEVPPDT
ASPH is thought to play an important role in calcium homeostasis. Alternative splicing of this gene results in five transcript variants which vary in protein translation, the coding of catalytic domains, and tissue expression. Variation among these transcripts impacts their functions which involve roles in the calcium storage and release process in the endoplasmic and sarcoplasmic reticulum as well as hydroxylation of aspartic acid and asparagine in epidermal growth factor-like domains of various proteins. This gene is thought to play an important role in calcium homeostasis. Alternative splicing of this gene results in five transcript variants which vary in protein translation, the coding of catalytic domains, and tissue expression. Variation among these transcripts impacts their functions which involve roles in the calcium storage and release process in the endoplasmic and sarcoplasmic reticulum as well as hydroxylation of aspartic acid and asparagine in epidermal growth factor-like domains of various proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A beta H-J-J; ASP beta-hydroxylase; Aspartate beta-hydroxylase; Aspartyl/asparaginyl beta-hydroxylase; aspartyl/asparaginyl-beta-hydroxylase; cardiac junctin; humbug; junctate; Peptide-aspartate beta-dioxygenase
Gene Aliases: AAH; ASPH; BAH; CASQ2BP1; FDLAB; HAAH; JCTN; junctin
UniProt ID: (Human) Q9H291
Entrez Gene ID: (Human) 444
Molecular Function: protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support