Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Transcriptional repressor. It inhibits the enhancer element of the AFP gene by binding to its AT-rich core sequence. Regulator of myoblasts differentiation through the binding to the AT-rich sequence of MYF6 promoter and promoter repression. Down-regulates the MUC5AC promoter in gastric cancer. Miura Y., J. Biol. Chem. 270:26840-26848(1995). Morinaga T., Mol. Cell. Biol. 11:6041-6049(1991). Martin J., Nature 432:988-994(2004).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-fetoprotein enhancer binding protein; Alpha-fetoprotein enhancer-binding protein; AT motif binding factor 1; AT motif-binding factor 1; AT-binding transcription factor 1; ZFH-3; zinc finger homeobox 3; Zinc finger homeobox protein 3; Zinc finger homeodomain protein 3
Gene Aliases: A230102L03Rik; ATBF1; ATBT; C16orf47; mKIAA4228; RGD1560268; WBP9; ZFHX3; ZNF927
UniProt ID: (Human) Q15911
Entrez Gene ID: (Human) 463, (Rat) 307829, (Mouse) 11906
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support