Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MKFKLHVNSA RQYKDLWNMS DDKPFLCTAP GCGQRFTNED HLAVHKHKHE MTLKFGPARN DSVIVADQTP TPTRFLKNCE EVGLFNELAS PFENEFKKAS EDDIKKMPLD LSPLATPIIR SKIEEPSVVE TTHQDSPLPH PESTTSDEKE VPLAQTAQPT SAIVRPASLQ VPNVLLTSSD SSVIIQQAVP SPTSSTVITQ APSSNRPIV
The transcription factor ATF-2 (also called CRE-BP1) binds to both AP-1 and CRE DNA response elements and is a member of the ATF/CREB family of leucine zipper proteins. ATF-2 interacts with a variety of viral oncoproteins and cellular tumor suppressors and is a target of the SAPK/JNK and p38 MAP kinase signaling pathways. Various forms of cellular stress, including genotoxic agents, inflammatory cytokines and UV irradiation, stimulate the transcriptional activity of ATF-2. Cellular stress activates ATF-2 by phosphorylation of Thr69 and Thr71. Both SAPK and p38 MAPK have been shown to phosphorylate ATF-2 at these sites in vitro and in cells transfected with ATF-2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating 2; Activating transcription factor 2; activating transcription factor 2 splice variant ATF2-var2; cAMP response element-binding protein CRE-BP1; cAMP responsive element binding protein 2, formerly; cAMP-dependent transcription factor ATF-2; CREB-2; Cyclic AMP-dependent transcription factor ATF-2; Cyclic AMP-responsive element-binding protein 2; HB16; histone acetyltransferase ATF2; MGC111558; OTTHUMP00000205345; OTTHUMP00000205347; OTTHUMP00000205348; OTTHUMP00000205350; OTTHUMP00000205352; OTTHUMP00000205353; OTTHUMP00000205354
Gene Aliases: ATF2; CRE-BP1; CREB-2; CREB2; CREBP1; HB16; TREB7
UniProt ID: (Human) P15336
Entrez Gene ID: (Human) 1386
Molecular Function:
basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support