Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QKEFVITSL KELGYVTLMC GDGTNDVGAL KHADVGVALL ANAPERVVER RRRPRDSPTL SNSGIRATSR TAKQRSGLPP SEEQPTSQRD RLSQVLRDLE DESTPIVKLG DASIAAPFTS KLSSIQCICH VIKQGRCTLV TTLQM (615-758 aa encoded by BC009302)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase type 13A1; cation transporting ATPase; cation-transporting ATPase 2; DKFZp761L1623; Endoplasmic reticulum P5A-ATPase; Endoplasmic reticulum transmembrane helix translocase; FLJ31858; FLJ41786; FLJ43873; FLJ90317; manganese-transporting ATPase 13A1; probable cation-transporting ATPase 13A1
Gene Aliases: ATP13A; ATP13A1; catp; CGI-152; Cgi152; KIAA1825
UniProt ID: (Human) Q9HD20, (Mouse) Q9EPE9
Entrez Gene ID: (Human) 57130, (Mouse) 170759, (Rat) 290673
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support