Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: KPLWGVRLRL RPCNLAHAET LVIEIRDKED SSWQLFTVQV QTEAIGEGSL EPSPQSQAED GRSQAAVGAV PEGAWKDTAQ LHKSEEA
Neurodegenerative disorders such as Parkinson and Alzheimer disease cause motor and cognitive dysfunction and belong to a heterogeneous group of common and disabling disorders. ATP13A2, otherwise known as PARK9, is a neuronal P-type ATPase gene underlying an autosomal recessive form of early-onset parkinsonism with pyramidal degeneration and dementia. ATP13A2 protein is located in the membrane of these lysosomes and is formed most strongly in the brain, especially in the substantia nigra, a brain region which is known to play a central role in Parkinson's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AT132 antibody; ATPase type 13A2; HSA9947 antibody; Polyamine-transporting ATPase 13A2; putative ATPase; RP1-37C10.4
Gene Aliases: ATP13A2; CLN12; HSA9947; KRPPD; PARK9
UniProt ID: (Human) Q9NQ11
Entrez Gene ID: (Human) 23400
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support