Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLSRVVLSAA ATAAPSLKNA AFLGPGVLQA TRTFHTGQPH LVPVPPLPEY GGKVRYGLIP EEFFQFLYPK TGVTGPYVLG TGLILYALSK EIYVISAETF TALSVLGVMV YGIKKYGPFV ADFADKLNEQ KLAQLEEAKQ ASIQHIQNAI DTEKSQQALV QKRHYLFDVQ RNNIAMALEV TYRERLYRVY KEVKNRLDYH ISVQNMMRRK EQEHMINWVE KHVVQSISTQ QEKETIAKCI ADLKLLAKKA QAQPVM
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene encodes the b subunit of the proton channel.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP synthase B chain, mitochondrial; ATP synthase F(0) complex subunit B1, mitochondrial; ATP synthase peripheral stalk-membrane subunit b; ATP synthase proton-transporting mitochondrial F(0) complex subunit B1; ATP synthase subunit b; ATP synthase subunit b, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b, isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit B1; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit B1; ATPase subunit b; cell proliferation-inducing protein 47; H+-ATP synthase subunit b
Gene Aliases: ATP5F1; ATP5PB; PIG47
UniProt ID: (Human) P24539
Entrez Gene ID: (Human) 515
Molecular Function:
ATP synthase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support