Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MILQRLFRFS SVIRSAVSVH LRRNIGVTAV AFNKELDPIQ KLFVDKIREY KSKRQTSGGP VDASSEYQQE LERELFKLKQ MFGNADMNTF PTFKFEDPKF EVIE
Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 77%.
Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. It is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, F0, which comprises the proton channel. The F1 complex consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled in a ratio of 3 alpha, 3 beta, and a single representative of the other 3. The F0 seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene encodes the F6 subunit of the F0 complex, required for F1 and F0 interactions. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP synthase peripheral stalk subunit F6; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F6; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F6; ATP synthase-coupling factor 6, mitochondrial; ATPase subunit F6; C ATP5; coupling factor 6; mitochondrial ATP synthase, coupling factor 6; mitochondrial ATP synthase, subunit F6; mitochondrial ATPase coupling factor 6; proliferation-inducing protein 36
Gene Aliases: ATP5; ATP5A; ATP5J; ATP5PF; ATPM; CF6; F6
UniProt ID: (Human) P18859
Entrez Gene ID: (Human) 522
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support