Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAQFVRNLV EKTPALVNAA VTYSKPRLAT FWYYAKVELV PPTPAEIPRA IQSLKKIANS AQTGSFKQLT VKEAVLNGLV ATEVLMWFYV GEIIGKRGII GYDV (1-103 aa encoded by BC015128)
UCK2 catalyzes the phosphorylation of uridine monophosphate to uridine diphosphate. This is the first step in the production of the pyrimidine nucleoside triphosphates required for RNA and DNA synthesis. In addition, an allele of this gene may play a role in mediating nonhumoral immunity to Hemophilus influenzae type B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP synthase g chain, mitochondrial; ATP synthase membrane subunit g; ATP synthase subunit g, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit G; ATP synthase, H+ transporting, mitochondrial F1F0, subunit g; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit G; ATPase subunit G; F1F0-ATP synthase g subunit; F1F0-type ATP synthase subunit g; F1Fo-ATP synthase complex Fo membrane domain g subunit
Gene Aliases: 4933437C06Rik; ATP5JG; ATP5L; ATP5MG
UniProt ID: (Human) O75964, (Rat) Q6PDU7, (Mouse) Q9CPQ8
Entrez Gene ID: (Human) 10632, (Rat) 300677, (Mouse) 27425
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support