Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: ISDLPAVSNP GLTQILVEEM LPNCEVVGVD WAEGLKSGPE EQPRDTASPV
Sequence homology: Guinea Pig: 77%; Human: 100%; Rabbit: 91%
Required for the assembly of the mitochondrial NADH:ubiquinone oxidoreductase complex (complex I). Involved in the assembly of the distal region of complex I. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP synthase subunit s-like protein; ATP5S-like; Distal membrane-arm assembly complex protein 2
Gene Aliases: ATP5SL; DMAC2
UniProt ID: (Human) Q9NW81
Entrez Gene ID: (Human) 55101
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support