Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
ATP6V0D2 is the subunit of the integral membrane V0 complex of vacuolar ATPase. Vacuolar ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells, thus providing most of the energy required for transport processes in the vacuolar system. ATP6V0D2 may play a role in coupling of proton transport and ATP hydrolysis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d2; FLJ38708; V-ATPase subunit d 2; V-type proton ATPase subunit d 2; Vacuolar proton pump subunit d 2
Gene Aliases: ATP6D2; ATP6V0D2; VMA6
UniProt ID: (Human) Q8N8Y2
Entrez Gene ID: (Human) 245972
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support