Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MALSDADVQK QIKHMMAFIE QEANEKAEEI DAKAEEEFNI EKGRLVQTQR LKIMEYYEKK EKQIEQQKKI QMSNLMNQAR LKVLRARDDL ITDLLNEAKQ RLSKVVKDTT RYQVLLDGLV LQGLYQLLEP RMIVRCRKQD FPLVKAAVQK AIPMYKIATK NDVDVQIDQE SYLPEDIAGG VEIYNGDRKI KVSNTLESRL DLIAQQMMPE VRGALFGANA NRKFLD
Subunit of the peripheral V1 complex of vacuolar ATPase essential for assembly or catalytic function. V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E1; H(+)-transporting two-sector ATPase, 31kDa subunit; H+-transporting ATP synthase chain E, vacuolar; p31; V-ATPase 31 kDa subunit; V-ATPase subunit E 1; V-ATPase, subunit E; V-type proton ATPase subunit E 1; Vacuolar proton pump subunit E 1
Gene Aliases: ATP6E; ATP6E2; ATP6V1E; ATP6V1E1; P31; Vma4
UniProt ID: (Human) P36543
Entrez Gene ID: (Human) 529
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support