Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTSQSQGIHQ LLQAEKRAKD KLEEAKKRKG KRLKQAKEEA MVEIDQYRMQ RDKEFRLKQS KIMGSQNNLS DEIEEQTLGK IQELNGHYNK YMESVMNQLL SMVCDMKPEI HVNYRATN
ATP6V1G3 (V-ATPase G3) is a catalytic subunit of the peripheral V1 complex of vacuolar ATPase (V-ATPase). V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, H+ transporting, lysosomal (vacuolar proton pump) subunit G3; ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G3; V-ATPase 13 kDa subunit 3; V-ATPase G subunit 3; V-ATPase G3 subunit; V-ATPase subunit G 3; V-type proton ATPase subunit G 3; vacuolar ATP synthase subunit G 3; vacuolar proton pump G subunit 3; Vacuolar proton pump subunit G 3; vacuolar proton pump, subunit G3
Gene Aliases: ATP6G3; ATP6V1G3; Vma10
UniProt ID: (Human) Q96LB4
Entrez Gene ID: (Human) 127124
Molecular Function:
ATP synthase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support