Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: IMTFKRCSIN GRIYGEVHDD LDQKTEITQE KEPVDFSVKS QADREFQFFD HHLMESIKMG DPKVHEFLRL LALCHTVMSE ENSAGELI
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase class I type 8B member 4; ATPase, class I, type 8B, member 4; P4-ATPase flippase complex alpha subunit ATP8B4; potential phospholipid-transporting ATPase IM; Probable phospholipid-transporting ATPase IM
Gene Aliases: ATP8B4; ATPIM; KIAA1939
UniProt ID: (Human) Q8TF62
Entrez Gene ID: (Human) 79895
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support