Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YSWVIRRDSK IPGTVVRSST IPEQLGRISY LLTDKTGTLT QNEMIFKRLH LGTVAYGLDS MDEVQSHIFS IYTQQSQDPP AQKGPTLTTK VRRTMSSRV
ATP9A, also named as ATPIIA and KIAA0611, belongs to the cation transport ATPase (P-type) family and Type IV subfamily. ATP9A catalyzes the reaction: ATP + H2O + phospholipid(In) = ADP + phosphate + phospholipid(Out).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase class II type 9A; ATPase type IV, phospholipid-transporting (P-type),(putative); ATPase, class II, type 9A; phospholipid-transporting ATPase IIA; Probable phospholipid-transporting ATPase IIA
Gene Aliases: ATP9A; ATPIIA; KIAA0611
UniProt ID: (Human) O75110
Entrez Gene ID: (Human) 10079
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support