Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Multiple transcript variants encoding different isoforms exist for this gene. One of the isoforms is a membrane-bound protein with sequence similarity to the mouse mahogany protein, a receptor involved in controlling obesity. The other isoform is a secreted protein involved in the initial immune cell clustering during inflammatory responses that may regulate the chemotactic activity of chemokines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: atrn {ECO:0000312|RGD:69063}; Attractin; attractin-2; DPPT-L; Mahogany homolog; mahogany protein; membrane attractin; MGC126754; Protein mahogany; Protein zitter
Gene Aliases: ATRN; AW558010; DPPT-L; KIAA0548; mahogany; Mg; MGCA; mKIAA0548
UniProt ID: (Human) O75882, (Rat) Q99J86, (Mouse) Q9WU60
Entrez Gene ID: (Human) 8455, (Rat) 83526, (Mouse) 11990
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support