Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATRNL1 (Attractin Like 1) is a Protein Coding gene. May play a role in melanocortin signaling pathways that regulate energy homeostasis. Gene Ontology (GO) annotations related to this gene include carbohydrate binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: attractin like 1; attractin-like 1; Attractin-like protein 1
Gene Aliases: AI504415; ALP; Atrnl; ATRNL1; Atrnlp; AW555641; bA338L11.1; bA454H24.1; KIAA0534; RGD1563485
UniProt ID: (Human) O60283, (Mouse) Q6A051
Entrez Gene ID: (Human) 26033, (Mouse) 226255, (Rat) 307992
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support