Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SLPLSFDKSE GKKRKNSSSS SKACKITKMP GMNSVHKKNP PSLLAPVPDP VNSTSSRQVG KNSSLALSQS SPSSISSPGH SRQKNTNRTG RIR
Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 86%.
ATXN7L1 (ataxin-7-like protein 1) is an 833 amino acid protein that contains one SCA7 domain. The ATXN7L1 gene is conserved in chimpanzee, canine, mouse, rat and chicken, and maps to human chromosome 7q22.3. Chromosome 7 is about 158 million bases long, encodes over 1,000 genes and makes up about 5%of the human genome. Chromosome 7 has been linked to osteogenesis imperfecta, Pendred syndrome, lissencephaly, citrullinemia and Shwachman Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterized by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ataxin 7-like 1; ataxin 7-like 4; Ataxin-7-like protein 1; Ataxin-7-like protein 4
Gene Aliases: ATXN7L1; ATXN7L4; KIAA1218
UniProt ID: (Human) Q9ULK2
Entrez Gene ID: (Human) 222255
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support