Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Histone H1 protein binds to linker DNA between nucleosomes forming the macromolecular structure known as the chromatin fiber. Histones H1 are necessary for the condensation of nucleosome chains into higher-order structured fibers. Acts also as a regulator of individual gene transcription through chromatin remodeling, nucleosome spacing and DNA methylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H1 histone family, member 4; histone 1, H1e; Histone H1.4; Histone H1b; Histone H1s-4
Gene Aliases: dJ221C16.5; H1-4; H1.4; H1E; H1F4; H1s-4; HIST1H1E
UniProt ID: (Human) P10412
Entrez Gene ID: (Human) 3008
Molecular Function:
histone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support