Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Component of adherens-type cell-cell and cell-matrix junctions. Lacks phosphoglucomutase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aciculin; PGM-RP; Phosphoglucomutase-like protein 5; Phosphoglucomutase-related protein
Gene Aliases: PGM5; PGMRP
UniProt ID: (Human) Q15124
Entrez Gene ID: (Human) 5239
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support