Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GRVHPNTRA NYLASPPLVI AYAIAGTIRI DFEKEPLGVN AKGQQVFLKD IWPTRDEIQA VERQYVIPGM FKEVYQKIET VNESWNALAT PSDKLFFWNS KSTYIKSPPF FENLTLDLQP PKSIVDAYVL LNLGDSVTTD HISPAGNIAR NSPAARYLTN RGLTPREFNS YGSRRGNDAV MARGTFANIR LLNRFLNKQA PQTIHLPSGE ILDVFDAAER YQQAGLPLIV LAGKEYGAGS SRDWAAKGPF LLGIKAVLAE SYERIHRSNL VGMGVIPLEY LPGENADALG LTGQERYTII IPENLKPQMK VQVKLDTGKT FQAVMRFDTD VELTYFLNGG ILNYMIRKMA K (540-890 aa encoded by BC018103)
Aconitase 1 (ACO1) also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). It plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes.Aconitase 1, also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). IREs are stem-loop structures found in the 5' UTR of ferritin mRNA, and in the 3' UTR of transferrin receptor mRNA. The iron-induced binding to the IRE results in repression of translation of ferritin mRNA, and inhibition of degradation of the otherwise rapidly degrading transferrin receptor mRNA. Thus, IREB1 plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aconitase; aconitase 1, soluble; aconitate hydratase, cytoplasmic; Citrate hydro-lyase; cytoplasmic aconitase; Cytoplasmic aconitate hydratase; cytosplasmic aconitase; epididymis luminal protein 60; Ferritin repressor protein; IRE-BP 1; Iron regulatory protein 1; iron-responsive element binding protein 1; Iron-responsive element-binding protein 1; IRP1; OTTHUMP00000021176; OTTHUMP00000021177; OTTHUMP00000045233; soluble aconitase
Gene Aliases: Aco-1; ACO1; Acon1; ACONS; AH; HEL60; IREB1; IREBP; IREBP1; IRP1
UniProt ID: (Human) P21399, (Rat) Q63270, (Mouse) P28271
Entrez Gene ID: (Human) 48, (Rat) 50655, (Mouse) 11428
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support