Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. In vertebrates there are three groups of actin isoforms: alpha, beta and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exists in most cell types as components of the cytoskeleton and as mediators of internal cell motility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACTB; actin; actin alpha 1; actin alpha cardiac; actin alpha cardiac 1; actin, alpha 2, smooth muscle, aorta; Actin, alpha cardiac muscle 1; Actin, alpha skeletal muscle; actin, alpha, cardiac 1; actin, alpha, vascular smooth muscle; Actin, aortic smooth muscle; actin, beta; Actin, cytoplasmic 1; Actin, cytoplasmic 2; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; Actin, gamma-enteric smooth muscle; actin-like protein; alpha actin 1; alpha actin 2; alpha-actin cardiac; Alpha-actin-1; Alpha-actin-2; Alpha-actin-3; Alpha-cardiac actin; alpha-sma; alpha-smooth muscle actin; alphac-actin; Atcb; B-actin; beta actin; beta cytoskeletal actin; Beta-actin; beta-actin FE-3; cardiac alpha actin; cardiac muscle alpha actin 1; Cell growth-inhibiting gene 46 protein; cytoplasmic beta-actin; cytoskeletal beta actin; cytoskeletal gamma-actin; deafness, autosomal dominant 20; deafness, autosomal dominant 26; epididymis luminal protein 176; Gamma-2-actin; Gamma-actin; gamma-enteric smooth muscle actin; nemaline myopathy type 3; pan actin; PS1TP5-binding protein 1; RP5-1068B5.2; skeletal alpha actin; skeletal muscle alpha-actin 1; smooth muscle alpha-actin; Smooth muscle gamma-actin; vascular smooth muscle alpha-actin
Gene Aliases: 0610041G09Rik; a-SMA; AA959943; AAT6; ACT; ACT-4; Act4; ACTA; Acta-2; ACTA1; ACTA2; ACTA3; ACTB; ACTC; Actc-1; ACTC1; ACTE; ACTG; ACTG1; ACTG2; ACTGE; actin; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; ACTVS; Actx; AL023024; alphaSMA; ASD5; ASMA; beta-actin; BRWS1; BRWS2; CFTD; CFTD1; CFTDM; CMD1R; CMH11; DFNA20; DFNA26; E430023M04Rik; E51; GIG46; HEL-176; LVNC4; MPFD; MYMY5; NEM1; NEM2; NEM3; PS1TP5BP1; SHPM; SMalphaA; SMGA; VSCM
UniProt ID: (Human) P68133, (Bovine) P68138, (Pig) P68137, (Bovine) P62739, (Human) P04108, (Horse) P60708, (Bovine) P60712, (Pig) Q6QAQ1, (Human) Q96B34, (Human) P68032, (Bovine) Q3ZC07, (Human) P63261, (Bovine) P14104, (Bovine) Q5E9B5, (Human) P63267, (Mouse) P68134, (Rat) P68136, (Rat) P62738, (Mouse) P62737, (Rat) P60711, (Mouse) P60710, (Rat) P68035, (Mouse) P68033, (Mouse) P63260, (Rat) P63259, (Mouse) P63268, (Rat) P63269
Entrez Gene ID: (Horse) 100065454, (Human) 58, (Bovine) 281592, (Pig) 100154254, (Pig) 733615, (Bovine) 515610, (Horse) 100062175, (Human) 59, (Horse) 100033878, (Bovine) 280979, (Pig) 414396, (Human) 60, (Human) 70, (Bovine) 533219, (Horse) 100057678, (Pig) 100152267, (Human) 71, (Bovine) 404122, (Pig) 397653, (Horse) 100050139, (Bovine) 281595, (Pig) 100520667, (Human) 72, (Horse) 100057561, (Mouse) 11459, (Rat) 29437, (Rat) 81633, (Mouse) 11475, (Rat) 81822, (Mouse) 11461, (Rat) 29275, (Mouse) 11464, (Mouse) 11465, (Rat) 287876, (Mouse) 11468, (Rat) 25365
Molecular Function:
actin and actin related protein
actin or actin-binding cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support