Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SPTPGSEGHS AAPDCPSCAL AALPKDVPNS QPEMVEAVKK HILNMLHLKK RPDVTQPVPK AALLNAIRKL HVGKVGENGY VEIEDDIGRR AEMNELMEQT SEIITFAESG TARKTLHFEI SKEGSDLSVV ERAEVWLFLK VPKANRTRTK VTIRLFQQQK HPQGSLDTGE EAEEVGLKGE RSELLLSEKV VDARKSTWHV FPVSSSIQRL LDQGKSSLDV RIACEQCQES GASLVLLGKK KKKEEEGEGK KKGGGEGGAG ADEEKEQSHR PFLMLQARQS EDHPHRRRRR GLECDGKVNI CCKKQFFVSF KDIGWNDWII APSGYHANYC EGECPSHIAG TSGSSLSFHS TVINHYRMRG HSPFANLKSC CVPTKLRPMS MLYYDDGQNI IKKDIQNMIV EECGCS
Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Activin A mutations are associated with fibrodysplasia ossificans progressive.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activin A; Activin beta-A chain; EDF; erythroid differentiation factor; Erythroid differentiation protein; follicle-stimulating hormone-releasing protein; FSH-releasing protein; Inhibin beta A chain; inhibin, beta A (activin A, activin AB alpha polypeptide); Inhibin, beta-1
Gene Aliases: EDF; FRP; INHBA
UniProt ID: (Human) P08476, (Rat) P18331, (Mouse) Q04998
Entrez Gene ID: (Human) 3624, (Rat) 29200, (Mouse) 16323
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support