Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Store vial at 4° C prior to restoration. For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Biotin, anti-Rabbit Serum as well as purified and partially purified Adenosine Deaminase [Calf Spleen]. Cross reactivity against Adenosine Deaminase from other sources may occur but has not been specifically determined.
Catalyzes the hydrolytic deamination of adenosine and 2-deoxyadenosine. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. May act as a positive regulator of T-cell coactivation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenosine aminohydrolase; Adenosine deaminase; RP11-61L14.5
Gene Aliases: ADA
UniProt ID: (Bovine) P56658
Entrez Gene ID: (Bovine) 280712
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support