Search Thermo Fisher Scientific
The MA1-7327 antibody reacts with Adenovirus hexon in viral and infected tissue samples.
MA1-7327 has been successfully used in ELISA and Immunofluorescence applications.
The MA1-7327 antibody was raised against Adenovirus hexon protein.
Hexon protein is a major coat protein of adenoviruses. Adenoviruses capsids have three principal protein components: the hexon, the penton, and the fiber. Hexon consists of three subunits together forming two major components of different morphological symmetry. A triangular top with three towers of density is superimposed on a more bulky pseudo hexagonal base.
Protein Aliases: Adeno Virus; AdV; HADV; HAdVC_gp08