Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total AK5.
This gene encodes a member of the adenylate kinase family, which is involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate groups among adenine nucleotides. This member is related to the UMP/CMP kinase of several species. It is located in the cytosol and expressed exclusively in brain. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adenylate kinase 6; Adenylate kinase isoenzyme 5; AK 5; ATP-AMP transphosphorylase 5; MGC33326
Gene Aliases: AK5; AK6
UniProt ID: (Human) Q9Y6K8, (Mouse) Q920P5
Entrez Gene ID: (Human) 26289, (Mouse) 229949
Molecular Function:
nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support