Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKLVSVALM YLGSLAFLGA DTARLDVASE FRKKWNKWAL SRGKRELRMS SSYPTGLADV KAGPAQTLIR PQDMKGASRS PEDSSPDAAR IRVKRYRQSM NNFQGLRSFG CRFGTCTVQK LAHQIYQFTD KDKDNVAPRS KISPQGYGRR RRRSLPEAGP GRTLVSSKPQ AHGAPAPPSG SAPHFL (1-185 aa encoded by B C015961)
Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterized by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintenance of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is upregulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AMpreproadrenomedullin; preproadrenomedullin; Pro-adrenomedullin; proadrenomedullin N-20 terminal peptide; ProAM-N20
Gene Aliases: ADM; AM; PAMP
UniProt ID: (Human) P35318
Entrez Gene ID: (Human) 133
Molecular Function: peptide hormone intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support