Search Thermo Fisher Scientific
Chromotek
FIGURE: 1 / 4
Anti-human IgE Nanobody for site-directed immobilization of human IgE on plates, surfaces and biosensors.
Affinity: Dissociation constant KD of 0.72 nm. Apparent affinity may be higher for full IgEs due to avidity effects (1 antibody captured by 2 Nanobodies).
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.