Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KDNGCFRSS GSLLNNAIKG GVEDEVTLSA YITIALLEIP LTVTHPVVRN ALFCLESAWK TAQEGDHGSH VYTKALLAYA FALAGNQDKR KEVLKSLNEE AVKKDNSVHW ERPQKPKAPV GHFYEPQAPS AEVEMTSYVL LAYLTAQPAP TSEDLTSATN IVKWITKQQN AQGGFSSTQD TVVALHALSK YGAATFTRTG KAAQVTIQSS GTFSSKFQVD NNNRLLLQQV SLPELPGEYS MKVTGEGCVY LQTSLKYNIL PEKEEFPFAL GVQTLPQTCD EPKAHTSFQI SLSVSYTGSR SASNMAIVDV KMVSGFIPLK PTVKMLERSN HVSRTEVSSN HVLIYLDKVS NQTLSLFFTV LQDVPVRDLK PAIVKVYDYY ETDEFAIAEY NAPCSKDLGN A (1075-1474 aa encoded by BC040071)
Alpha 2-Macroglobulin (a2M), is a approximately 720-kDa homotetrameric glycoprotein composed of four identical approximately 180 kDa subunits. aAlpha 2M shares with other alpha-macroglobulins, like the complement components C3 and C4 and the pregnancy zone protein PZP, an extraordinary binding capacity for a variety of ligands. This allows the alpha-macroglobulins to serve as humoral defense barriers against foreign peptides in the plasma. Alpha 2M interacts and captures virtually any proteinase, often referred to as a panprotease inhibitor. In the brain of Alzheimer's disease (AD) patients, Alpha 2M also has been localized to diffuse amyloid plaques, supporting an important role for alpha 2M in AD etiopathology.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha 2 M; Alpha-2-M; Alpha-2-M (A2M); Alpha-2-macroglobulin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5; DKFZp779B086; S863 7
Gene Aliases: A2M; A2MD; CPAMD5; FWP007; S863-7
UniProt ID: (Human) P01023
Entrez Gene ID: (Human) 2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support