Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: SEMDNYEPG VYTEKVLEAT KLLSNTVMPR FTEQVEAAVE ALSSDPAQPM DENEFIDASR LVYDGIRDIR KAVLMIRTPE ELDDSDFETE DFDVRSRTSV QTEDDQLIAG QSARAIMAQL PQEQKAKIAE QVASFQEEKS KLDAEVSKWD DSGNDIIVLA KQMCMIMMEM TDFTRGKGPL KNTSDVISAA KKIAEAGSRM DKLGRTIADH CPDSACKQDL LAYLQRIALY CHQLNICSKV KAEVQNLGGE LVVSGVDSAM SLIQAAKNLM NAVVQTVKAS YVASTKYQKS QGMASLNLPA VSWKMKAPEK KPLVKREKQD ETQTKIKRAS QKKHVNPVQA LSEFKAMDSI (188-536 aa encoded by BC031262)
Associates with the cytoplasmic domain of a variety of cadherins. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties. Can associate with both E- and N-cadherins. Originally believed to be a stable component of E-cadherin/catenin adhesion complexes and to mediate the linkage of cadherins to the actin cytoskeleton at adherens junctions. In contrast, cortical actin was found to be much more dynamic than E-cadherin/catenin complexes and CTNNA1 was shown not to bind to F-actin when assembled in the complex suggesting a different linkage between actin and adherence junctions components. The homodimeric form may regulate actin filament assembly and inhibit actin branching by competing with the Arp2/3 complex for binding to actin filaments. May play a crucial role in cell differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 102 kDa cadherin-associated protein; alpha E catenin; Alpha E-catenin; alpha(E)-catenin; alpha-E-catenin; cadherin associated protein; Cadherin-associated protein; CAP102; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin alpha 1; Catenin alpha-1; Renal carcinoma antigen NY-REN-13
Gene Aliases: 2010010M04Rik; AA517462; AI988031; CAP102; Catna1; CTNNA1; MDPT2
UniProt ID: (Human) P35221, (Mouse) P26231
Entrez Gene ID: (Human) 1495, (Mouse) 12385
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support