Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
AA Sequence of recombinant protein: SVRVEQVVKP PQNKTESENT SDKPKRKKKG GKNGKNRRNR KKKNPCNAEF QNFCIHGECK YIEHLEAVTC KCQQEYFGER CGEKSMKTHS MIDSSLSK.
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Amphiregulin. Anti-Human Amphiregulin-specific antibody was purified by affinity chromatography and then biotinylated.
Sandwich ELISA: To detect Human Amphiregulin by sandwich ELISA (using 100 µL/well), a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Amphiregulin (500-P322) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human Amphiregulin.
Western Blot: To detect Human Amphiregulin by Western Blot analysis, this antibody can be used at a concentration of 0.1-0.2 µg/mL. When used in conjunction with compatible development reagents, the detection limit for Recombinant Human Amphiregulin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
500-P322BT-1MG will be provided as 2 x 500 µg
Amphiregulin (AR) binds to EGF-Receptor (EGFR) with lower affinity than EGF. The mature secreted form AR is an 84-amino acid residue glycosylated polypeptide growth regulator, which is generated by proteolytic processing of a 252-amino acid transmembrane precursor. Seven different polypeptide ligands, which derive from distinct genes, are capable of binding to the extracellular domain of EGFR. These ligands include EGF, TGFalpha, AR, HB-EGF, cripto-1, epiregulin, and betacellulin. All of these growth factors contain a characteristic EGF-like domain which is defined by 6 evenly spaced cysteine residues that generate 3 loops through the formation of disulfide bonds. AR protein has been localized by immunohistochemistry to the epithelium of the colon, stomach, pancreas, breast, and placenta. AR is reportedly overexpressed in human cancers of breast, colon, stomach, and pancreas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amphiregulin; amphiregulin B; AR; Colorectum cell-derived growth factor; CRDGF; MGC13647; schwannoma-derived growth factor
Gene Aliases: AR; AREG; AREGB; CRDGF; SDGF
UniProt ID: (Human) P15514
Entrez Gene ID: (Human) 374
Molecular Function:
growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support