Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Angiopoietin-like protein 4 (ANGPTL4), is a secreted protein selectively expressed in adipose tissue, liver, and placenta1-3 that plays a variety of roles in vivo, ranging from adipogenesis to angiogenesis to carcinogenesis. The expression of ANGPTL4 is also under nutritional and hormonal control. ANGPTL4 has been observed to induce a strong pro-angiogenic response independent of vascular endothelial growth factor (VEGF), and its expression has been described in hypoxic human tissues as well as a variety of cancers, including liposarcoma, hepatocellular carcinoma, and conventional renal cell carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 425O18-1; Angiopoietin-like protein 4; Angiopoietin-related protein 4; ANGPTL2; Fasting-i; Fasting-induced adipose factor; fibrinogen/angiopoietin-related protein; Hepatic fibrinogen/angiopoietin-related protein; HFARP; major histocompatibility complex region NG27; Secreted protein Bk89
Gene Aliases: Angptl4; Arp4; Bk89; Farp; Fiaf; Hfarp; Ng27; Pgar; Pgarg; Pp1158
UniProt ID: (Mouse) Q9Z1P8
Entrez Gene ID: (Mouse) 57875
Molecular Function:
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support