Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total APOA2.
At least 9 distinct polymorphic forms of apolipoproteins are known. The apolipoproteins act as stabilizers of the intact lipoprotein particles. Quantitative measurements of HDL, LDL, and VLDL particles of human serum are often used to estimate an individual's relative risk of coronary heart disease. In addition, quantitative immunological measurements of certain apolipoproteins ( especially A-1 and B) have been suggested to be more accurate estimators of coronary heart disease that measurements of lipoprotein particles (especially HDL and LDL). The apolipoproteins are derived from human plasma by density gradient ultracentrifugation and HPLC.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APO A2; Apo-AII; Apolipoprotein; Apolipoprotein A-II; Apolipoprotein A2; Apolipoprotein AII
Gene Aliases: Alp-2; Apo-AII; Apoa-2; ApoA-II; APOA2; APOAII; Hdl-1
UniProt ID: (Human) P02652, (Rat) P04638, (Mouse) P09813
Entrez Gene ID: (Human) 336, (Rat) 25649, (Mouse) 11807
Molecular Function:
apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support