Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: Rat liver, Rat plasma
Immunogen sequence: MGTRLLPALF LVLLVLGFEV QGTQQPQQDE MPSPTFLTQV KESLSSYWES AKTAAQNLYE KTYLPAVDEK LRDLYSKSTA AMSTYTGIFT DQVLSVLKGE E
Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein C gene family encodes four homologous proteins designated apoC-I to -IV, which specifically modulate the metabolism of triglyceride-rich lipoproteins. The human apoC-I gene maps to chromosome 19q13.2 and is expressed primarily in the liver where it is activated when monocytes differentiate into macrophages. The human apoC-II gene maps to chromosome 19q13.2 and encodes a 79 amino acid single chain protein that is a necessary cofactor for the activation of lipoprotein lipase, the enzyme that hydrolyzes triglycerides in plasma and transfers the fatty acids to tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APC 2; APO C2; Apo CII; Apo-CII; APOC 2; ApoC II; APOC2 protein; ApoCII; Apolipoprotein C II; Apolipoprotein C-II; Apolipoprotein C2; Apolipoprotein CII; ApolipoproteinCII; MGC75082
Gene Aliases: APC2; APO-CII; APOC-II; APOC2; RGD1560725
UniProt ID: (Human) P02655, (Rat) G3V8D4
Entrez Gene ID: (Human) 344, (Rat) 292697
Molecular Function: apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support