Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: VGATVGTATY QLLVALHHPE GPEPAQDLVS AQHKASELET PASAQMLECK
Sequence homology: Cow: 75%; Human: 100%; Rabbit: 75%; Rat: 79%
AQP10 is a member of the aquaglyceroporin family of integral membrane proteins. Members of this family function as water-permeable channels in the epithelia of organs that absorb and excrete water. AQP10 was shown to function as a water-selective channel, and could also permeate neutral solutes such as glycerol and urea. This gene encodes a member of the aquaglyceroporin family of integral membrane proteins. Members of this family function as water-permeable channels in the epithelia of organs that absorb and excrete water. This protein was shown to function as a water-selective channel, and could also permeate neutral solutes such as glycerol and urea.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AQP-10; Aquaglyceroporin-10; Aquaporin-10; Small intestine aquaporin
Gene Aliases: AQP10; AQPA_HUMAN
UniProt ID: (Human) Q96PS8
Entrez Gene ID: (Human) 89872
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support