Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
This antibody has been tested in direct-ELISA
1-aminocyclopropane-1-carboxylate synthase (ACS) enzymes catalyze the conversion of S-adenosyl-L-methionine (SAM) into 1-aminocyclopropane-1-carboxylate (ACC), a direct precursor of ethylene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-aminocyclopropane-1-carboxylate synthase 8; ACC synthase 8; S-adenosyl-L-methionine methylthioadenosine-lyase 8
Gene Aliases: 1-amino-cyclopropane-1-carboxylate synthase 8; ACS8; AT4G37770; T28I19.50; T28I19_50
UniProt ID: (Arabidopsis) Q9T065
Entrez Gene ID: (Arabidopsis) 829933
Molecular Function:
transaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support