Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
ACTR5 may be involved in transcription regulation via its interaction with the INO80 complex, a chromatin remodeling complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin-related protein 5; ARP5 actin related protein 5 homolog; ARP5 actin-related protein 5 homolog; hARP5; INO80 complex subunit M; Sarcoma antigen NY-SAR-16
Gene Aliases: AA545173; ACTR5; ARP5; B430109J19Rik; INO80M
UniProt ID: (Human) Q9H9F9
Entrez Gene ID: (Pig) 100156550, (Human) 79913, (Mouse) 109275, (Rat) 362258
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support