Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Plasmodesmal-associated membrane beta-1,3-glucanase involved in plasmodesmal callose degradation and functions in the gating of plasmodesmata.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (1->3)-beta-glucan endohydrolase 10; (1->3)-beta-glucanase 10; 3-beta-glucosidase 10-beta-glucan endohydrolase 10; AtBG_ppap; Beta-1,3-endoglucanase 10; Beta-1,3-glucanase 10; Glucan endo-1; Glucan endo-1,3-beta-glucosidase 10; Putative plasmodesmal associated protein
Gene Aliases: 3-GLUCANASE_PUTATIVE; ARABIDOPSIS THALIANA BETA-1; AT5G42100; ATBG_PAP; ATBG_PPAP; beta-1; MJC20.21; MJC20_21
UniProt ID: (Arabidopsis) Q9FHX5
Entrez Gene ID: (Arabidopsis) 834215
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support