Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Immunogen sequence: KLIGEELAQL KEQRVHKTDL ERVLEANDGS GMLDEDEEDL QRALALSRQE IDMEDEEADL RRAIQLSMQG SSRNISQDMT QTSGTNLTSE ELRKRREAYF EKQQQKQQQQ QQQQQQGDLS GQSSHPCERP ATSSGALGSD LGDAMSEEDM LQAAVTMSLE TVRNDLKTEG KK
Machado-Joseph disease is an autosomal dominant neurologic disorder, and is now known to be the same as previously described spinocerebellar ataxia-3. MJD protein (Ataxin-3) contains (CAG)n repeats in the coding region, and the expansion of these repeats from the normal 13-36 to 68-79 is the cause of Machado-Joseph disease. There is a negative correlation between the age of onset and CAG repeat numbers. This protein interacts with key regulators (CBP, p300 and PCAF) of transcription and represses transcription, and also acts as a histone-binding protein that regulates transcription. MJD is a deubiquitinating enzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ataxin-3; ataxin3; Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3) homolog; Machado-Joseph disease protein 1 homolog; spin
Gene Aliases: 2210008M02Rik; AI463012; AI647473; ataxin-3; ATX3; Atxn3; Mjd; MJD1; Rsca3; Sca3
UniProt ID: (Rat) O35815, (Mouse) Q9CVD2
Entrez Gene ID: (Rat) 60331, (Mouse) 110616
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support