Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The Aurora gene, a serine-threonine kinase, orginally discovered in Drosophila, has been found to be responsible for a chromosomal segregation defect. The mammalian homologues of Drosophila Aurora have been termed ARK1 (Aurora-related kinase 1) and ARK2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AIM-1; ARK-1; ARK-2; Aurora 1; Aurora 2; aurora A; aurora B; aurora family kinase 1; Aurora kinase A; Aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; Aurora- and IPL1-like midbody-associated protein 1; aurora-1; aurora-A; aurora-B; aurora-related kinase 1; aurora-related kinase 2; aurora/IPL1-like kinase; Aurora/IPL1-related kinase 1; Aurora/IPL1-related kinase 2; Breast tumor-amplified kinase; Ipl1- and aurora-related kinase 1; protein phosphatase 1, regulatory subunit 47; protein phosphatase 1, regulatory subunit 48; ratAurA; serine/threonine kinase 12; serine/threonine kinase 5; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine protein kinase 6; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 15; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase Ayk1; STK-1
Gene Aliases: AIK; AIK2; AIM-1; AIM1; AIRK1; AIRK2; AL022959; ARK-1; ARK-2; ARK1; ARK2; AU019385; AURA; AurB; AURKA; AURKB; aurkb-sv1; aurkb-sv2; Aurora-A; AW539821; AYK1; BTAK; IAK; IAK1; IPL1; PPP1R47; PPP1R48; STK-1; STK1; STK12; STK15; STK5; STK6; STK7
UniProt ID: (Human) O14965, (Human) Q96GD4, (Mouse) P97477, (Mouse) O70126, (Rat) O55099
Entrez Gene ID: (Dog) 485940, (Horse) 100050161, (Human) 6790, (Human) 9212, (Dog) 479492, (Horse) 100073050, (Mouse) 20878, (Rat) 261730, (Mouse) 20877, (Rat) 114592
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support