Search Thermo Fisher Scientific
Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/ml.
Avi tag is a biotin-acceptor peptide, GLNDIFEAQKIEWHE. The 15-residue peptide served as a substrate mimic for biotin ligase (BirA), which usually recognizes the much larger protein domain. Avi Tag Antibody is a useful tool in analysis of Avi fusion proteins.
The Avi tag is a biotin-acceptor peptide1, GLNDIFEAQKIEWHE. The 15-residue peptide served as a substrate mimic for biotin ligase (BirA), which usually recognizes the much larger protein domain.
Protein Aliases: Avi; BirA