Product References
A FoxA2+ long-term stem cell population is necessary for growth plate cartilage regeneration after injury.
Nature communications
Muruganandan S,Pierce R,Teguh DA,Perez RF,Bell N,Nguyen B,Hohl K,Snyder BD,Grinstaff MW,Alberico H,Woods D,Kong Y,Sima C,Bhagat S,Ho K,Rosen V,Gamer L,Ionescu AM
B35138 was used in Immunohistochemistry to show that when the growth plate is injured, FoxA2+ cells expand in response to trauma, and produce physeal cartilage for growth plate tissue regeneration.
Fri May 06 00:00:00 EDT 2022
Interrogation of Dystrophin and Dystroglycan Complex Protein Turnover After Exon Skipping Therapy.
Journal of neuromuscular diseases
Novak JS,Spathis R,Dang UJ,Fiorillo AA,Hindupur R,Tully CB,Mázala DAG,Canessa E,Brown KJ,Partridge TA,Hathout Y,Nagaraju K
B35138 was used in Immunohistochemistry to demonstrate the use of targeted mass spectrometry to evaluate the suitability and functionality of restored dystrophin isoforms in the context of disease and propose its use to optimize alternative gene correction strategies in development for DMD.
Fri Dec 17 00:00:00 EST 2021
Interrogation of Dystrophin and Dystroglycan Complex Protein Turnover After Exon Skipping Therapy.
Journal of neuromuscular diseases
Novak JS,Spathis R,Dang UJ,Fiorillo AA,Hindupur R,Tully CB,Mázala DAG,Canessa E,Brown KJ,Partridge TA,Hathout Y,Nagaraju K
B35138 was used in Immunohistochemistry to demonstrate the use of targeted mass spectrometry to evaluate the suitability and functionality of restored dystrophin isoforms in the context of disease and propose its use to optimize alternative gene correction strategies in development for DMD.
Fri Dec 17 00:00:00 EST 2021
Momordica charantia polysaccharides modulate the differentiation of neural stem cells via SIRT1/Β-catenin axis in cerebral ischemia/reperfusion.
Stem cell research & therapy
Hu Z,Li F,Zhou X,Zhang F,Huang L,Gu B,Shen J,Qi S
Published figure using BrdU monoclonal antibody (Product # B35138) in Immunohistochemistry
Mon Nov 16 00:00:00 EST 2020
Momordica charantia polysaccharides modulate the differentiation of neural stem cells via SIRT1/Β-catenin axis in cerebral ischemia/reperfusion.
Stem cell research & therapy
Hu Z,Li F,Zhou X,Zhang F,Huang L,Gu B,Shen J,Qi S
Published figure using BrdU monoclonal antibody (Product # B35138) in Immunohistochemistry
Mon Nov 16 00:00:00 EST 2020
TGF-β-driven muscle degeneration and failed regeneration underlie disease onset in a DMD mouse model.
JCI insight
Mázala DA,Novak JS,Hogarth MW,Nearing M,Adusumalli P,Tully CB,Habib NF,Gordish-Dressman H,Chen YW,Jaiswal JK,Partridge TA
B35138 was used in Immunohistochemistry-immunofluorescence to provide in vivo evidence of early-stage deficit in regenerative myogenesis in D2-mdx mice and implicates TGF-β as a major component of a pathogenic positive feedback loop in this model, identifying this feedback loop as a therapeutic target.
Thu Mar 26 00:00:00 EDT 2020
Pharmacologic ATM but not ATR kinase inhibition abrogates p21-dependent G1 arrest and promotes gastrointestinal syndrome after total body irradiation.
Scientific reports
Vendetti FP,Leibowitz BJ,Barnes J,Schamus S,Kiesel BF,Abberbock S,Conrads T,Clump DA,Cadogan E,O'Connor MJ,Yu J,Beumer JH,Bakkenist CJ
B35138 was used in immunohistochemistry - paraffin section to discover that ATM kinase activity is essential for p21-dependent and p21-independent mechanisms that radioprotect intestinal crypts
Wed Feb 01 00:00:00 EST 2017
Context-Dependent Effects of Amplified MAPK Signaling during Lung Adenocarcinoma Initiation and Progression.
Cell reports
Cicchini M,Buza EL,Sagal KM,Gudiel AA,Durham AC,Feldser DM
Published figure using BrdU monoclonal antibody (Product # B35138) in Immunofluorescence
Tue Feb 21 00:00:00 EST 2017
Myoblasts and macrophages are required for therapeutic morpholino antisense oligonucleotide delivery to dystrophic muscle.
Nature communications
Novak JS,Hogarth MW,Boehler JF,Nearing M,Vila MC,Heredia R,Fiorillo AA,Zhang A,Hathout Y,Hoffman EP,Jaiswal JK,Nagaraju K,Cirak S,Partridge TA
B35138 was used in Immunohistochemistry to show that robust morpholino (PMO) uptake and efficient production of dystrophin following PMO administration coincide with areas of myofiber regeneration and inflammation.
Mon Oct 16 00:00:00 EDT 2017
Chronic Exposure to the Fusarium Mycotoxin Deoxynivalenol: Impact on Performance, Immune Organ, and Intestinal Integrity of Slow-Growing Chickens.
Toxins
Chen SS,Li YH,Lin MF
B35138 was used in Immunohistochemistry to investigate the long-term effects of deoxynivalenol consumption on avian growth performance, on the proliferation, apoptosis, and DNA damage of spleen cells, and on intestinal integrity.
Fri Oct 20 00:00:00 EDT 2017
Effect of genetic background on the dystrophic phenotype in mdx mice.
Human molecular genetics
Coley WD,Bogdanik L,Vila MC,Yu Q,Van Der Meulen JH,Rayavarapu S,Novak JS,Nearing M,Quinn JL,Saunders A,Dolan C,Andrews W,Lammert C,Austin A,Partridge TA,Cox GA,Lutz C,Nagaraju K
Published figure using BrdU monoclonal antibody (Product # B35138) in Immunofluorescence
Fri Jan 01 00:00:00 EST 2016
Muscular dystrophy in the mdx mouse is a severe myopathy compounded by hypotrophy, hypertrophy and hyperplasia.
Skeletal muscle
Duddy W,Duguez S,Johnston H,Cohen TV,Phadke A,Gordish-Dressman H,Nagaraju K,Gnocchi V,Low S,Partridge T
Published figure using BrdU monoclonal antibody (Product # B35138) in Immunofluorescence
Tue May 19 00:00:00 EDT 2015
Annexin A1 Deficiency does not Affect Myofiber Repair but Delays Regeneration of Injured Muscles.
Scientific reports
Leikina E,Defour A,Melikov K,Van der Meulen JH,Nagaraju K,Bhuvanendran S,Gebert C,Pfeifer K,Chernomordik LV,Jaiswal JK
Published figure using BrdU monoclonal antibody (Product # B35138) in Immunofluorescence
Tue Dec 15 00:00:00 EST 2015
Delta-9-tetrahydrocannabinol disrupts hippocampal neuroplasticity and neurogenesis in trained, but not untrained adolescent Sprague-Dawley rats.
Brain research
Steel RW,Miller JH,Sim DA,Day DJ
B35138 was used in immunohistochemistry - paraffin section to identify effects of Delta(9)-tetrahydrocannabinol on the early induction of adaptive processes that support learning
Sat Feb 22 00:00:00 EST 2014
p27 is regulated independently of Skp2 in the absence of Cdk2.
Biochimica et biophysica acta
Kotoshiba S,Gopinathan L,Pfeiffenberger E,Rahim A,Vardy LA,Nakayama K,Nakayama KI,Kaldis P
Published figure using BrdU monoclonal antibody (Product # B35138) in Immunohistochemistry
Sat Feb 01 00:00:00 EST 2014
A novel transgenic mouse model of growth plate dysplasia reveals that decreased chondrocyte proliferation due to chronic ER stress is a key factor in reduced bone growth.
Disease models & mechanisms
Gualeni B,Rajpar MH,Kellogg A,Bell PA,Arvan P,Boot-Handford RP,Briggs MD
B35138 was used in immunohistochemistry - paraffin section to elucidate decreased chondrocyte proliferation caused by chronic ER stress as a key factor in reduced bone growth in a novel transgenic mouse model of growth plate dysplasia
Fri Nov 01 00:00:00 EDT 2013