Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Beta-1,3-N-acetylgalactosaminyltransferase active in synthesizing a unique carbohydrate structure, GalNAc-beta-1-3GlcNAc, on N- and O-glycans. Has no galactose nor galactosaminyl transferase activity toward any acceptor substrate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1,3-GalNAc-T2; Beta-1,3-N-acetylgalactosaminyltransferase II; UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 2; UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 2
Gene Aliases: B3GalNAc-T2; B3GALNT2; MDDGA11
UniProt ID: (Human) Q8NCR0
Entrez Gene ID: (Human) 148789
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support