Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: AEKIFKVSLG IRRLHLEDVY VGICLAKLRI DPVPPPNEFV FNHWRVSYSS CKYSHLITSH QFQPSELIKY WNHLQQNKHN ACANAAKEKA GRYRHRKLH
This gene is a member of the beta-1,3-galactosyltransferase (beta3GalT) gene family. This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine). The beta3GalT genes are distantly related to the Drosophila Brainiac gene and have the protein coding sequence contained in a single exon. The beta3GalT proteins also contain conserved sequences not found in the beta4GalT or alpha3GalT proteins. The carbohydrate chains synthesized by these enzymes are designated as type 1, whereas beta4GalT enzymes synthesize type 2 carbohydrate chains. The ratio of type 1:type 2 chains changes during embryogenesis. By sequence similarity, the beta3GalT genes fall into at least two groups: beta3GalT4 and 4 other beta3GalT genes (beta3GalT1-3, beta3GalT5). This gene encodes a protein that functions in N-linked glycoprotein glycosylation and shows strict donor substrate specificity for UDP-galactose.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1,3-galactosyltransferase 2; Beta-1,3-GalTase 2; beta-3-galt2; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase 2; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 2; UDP-galactose:2-acetamido-2-deoxy-D-glucose 3beta-galactosyltransferase 2
Gene Aliases: B3GALT2; beta3Gal-T2; BETA3GALT2; GLCT2
UniProt ID: (Human) O43825
Entrez Gene ID: (Human) 8707
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support