Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: VHKDEGSDPR RETPPGADPR EYCTSDRDIV EVVRTEYVYT RPPPWSDTLP TIHVVTPTYS RPVQKAELTR MANTLLHVPN LHWLVVEDAP RRTPLTARLL RDTGLNYTHL HVETPRNYKL RGDARDPRIP RGTMQRNLAL RWLRETFPRN SSQPGVVYFA DDDNTYSLEL FEEMRSTRRV SVWPVAFVGG LRYEAPRVNG AGKVVGWKTV FDPHRPFAID MAGFAVNLRL ILQRSQAYFK LRGVKGGYQE SSLLRELVTL NDLEPKAANC TKILVWHTRT EKPVLVNEGK KGFTDPSVEI; Positive Samples: SW480, HT-1080, BT-474; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein
The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1,3-glucuronyltransferase 1; CD57; Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1; GlcAT-P; GLCATP; GlcUAT-P; GLCUATP; Glucuronosyltransferase P; LEU7 antigen; NK-1; NK1; UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
Gene Aliases: B3GAT1; CD57; GLCATP; GLCUATP; HNK1; LEU7; NK-1; NK1
UniProt ID: (Human) Q9P2W7
Entrez Gene ID: (Human) 27087
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support