Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CKVQEQPPA IPEALAWPTP PTRPAPAPCH ANTSMVTHPD FATQPQHVQN FLLYRHYRHF PLLQDVPPSK CAQPVFLLLV IKSSPSNYVR RELLRRTWGR ERKVRGLQLR LLFLVGTASN PHEARKVNRL LELEAQTHGD ILQWDFHDSF FNLTLKQVLF LQWQETRCAN ASFVLNGDDD VFAHTDNMVF YLQDHDPGRH LFVGQLIQNV GPIRAFWSKY YVPEVVTQNE RYPPYCGGGG FLLSRFTAAA LRRAAHVLDI FPIDDVFLGM CLELEGLKPA SHSGIRTSGV RAPSQHLSSF DPCFYRDLLL VHRFLPYEML LMWDALNQPN LTCGNQTQIY (34-372 aa encoded by BC066876)
This gene encodes a member of the beta-1, 3-N-acetylglucosaminyltransferase family. This enzyme is a type II transmembrane protein and contains a signal anchor that is not cleaved. It prefers the substrates of lacto-N-tetraose and lacto-N-neotetraose, and is involved in the biosynthesis of poly-N-acetyllactosamine chains and the biosynthesis of the backbone structure of dimeric sialyl Lewis a. It plays dominant roles in L-selectin ligand biosynthesis, lymphocyte homing and lymphocyte trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase; Beta-1,3-galactosyltransferase 8; Beta-1,3-GalTase 8; beta-1,3-Gn-T3; beta-1,3-N-acetylglucosaminyltransferase 3; beta-1,3-N-acetylglucosaminyltransferase bGnT-3; Beta-3-Gx-T8; beta3GalT8; beta3Gn-T3; BGnT-3; core 1 beta-1,3-N-acetylglucosaminyltransferase; Core 1 extending beta-1,3-N-acetylglucosaminyltransferase; Core1-beta3GlcNAcT; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 3; putative type II membrane protein; Transmembrane protein 3; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 8; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 8; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3
Gene Aliases: 2210008L19Rik; B3GAL-T8; B3GALT8; B3GN-T3; B3GNT-3; B3GNT3; beta3Gn-T3; HP10328; TMEM3; UNQ637/PRO1266
UniProt ID: (Human) Q9Y2A9, (Mouse) Q5JCS9
Entrez Gene ID: (Human) 10331, (Mouse) 72297
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support