Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLPPQASAAH QGRGGRSGLL PKGPAMLCRL CWLVSYSLAV LLLGCLLFLR KAAKPAGDPT AHQPFWAPPT PRHSRCPPNH TVSSASLSLP SRHRLFLTYR HCRNFSILLE PSGCSKDTFL LLAIKSQPGH VERRAAIRST WGRVGGWARG RQLKLVFLLG VAGSAPPAQL LAYESREFDD ILQWDFTEDF FNLTLKELHL QRWVVAACPQ AHFMLKGDDD VFVHVPNVLE FLDGWDPAQD LLVGDVIRQA LPNRNTKVKY FIPPSMYRAT HYPPYAGGGG YVMSRATVRR LQAIMEDAEL FPIDDVFVGM CLRRLGLSPM HHAGFKTFGI RRPLDPLDPC LYRGLLLVHR LSPLEMWTMW ALVTDEGLKC AAGPIPQR
This gene encodes a member of the beta-1,3-N-acetylglucosaminyltransferase protein family. The encoded enzyme is involved in the biosynthesis of poly-N-acetyllactosamine chains and prefers lacto-N-neotetraose as a substrate. It is a type II transmembrane protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-1,3-Gn-T4; beta-1,3-N-acetylglucosaminyltransferase 4; beta-1,3-N-acetylglucosaminyltransferase bGn-T4; BGnT-4; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 4; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
Gene Aliases: B3GN-T4; B3GNT4; beta3Gn-T4; UNQ1898/PRO4344
UniProt ID: (Human) Q9C0J1
Entrez Gene ID: (Human) 79369
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support