Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PLLWDAPAK CAGGRGVFLL LAVKSAPEHY ERRELIRRTW GQERSYGGRP VRRLFLLGTP GPEDEARAER LAELVALEAR EHGDVLQWAF ADTFLNLTLK HLHLLDWLAA RCPHARFLLS GDDDVFVHTA NVVRFLQAQP PGRHLFSGQL MEGSVPIRDS WSKYFVPPQL FPGSAYPVYC SGGGFLLSGP TARALRAAAR HTPLFPIDDA YMGMCLERAG LAPSGHEGIR PFGVQLPGAQ QSS (102-343 aa encoded by BC103910)
The protein encoded by this gene is a beta-1,3-N-acetylglucosaminyltransferase that adds an N-acetylglucosamine moiety to N-acetylgalactosamine-modified serine or threonine. The encoded enzyme is responsible for creating the core 3 structure of O-glycans, which are important components of mucin-type glycoproteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acetylgalactosaminyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase; beta-1,3-Gn-T6; beta-1,3-N-acetylglucosaminyltransferase 6; beta-1,3-N-acetylglucosaminyltransferase protein; beta3Gn-T6; BGnT-6; Core 3 synthase; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Gene Aliases: B3Gn-T6; B3GNT6; BC039789; beta-1,3-Gn-T6; beta3Gn-T6; BGnT-6; Core-3; Core3
UniProt ID: (Human) Q6ZMB0, (Mouse) Q3USF0
Entrez Gene ID: (Human) 192134, (Mouse) 272411
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support