Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PGQFLQEPP PPTLEPQKAQ KPNGQLVNPN NFWKNPKDVA APTPMASQGP QAWDVTTTNC SANINLTHQP WFQVLEPQFR QFLFYRHCRY FPMLLNHPEK CRGDVYLLVV VKSVITQHDR REAIRQTWGR ERQSA (31-164 aa encoded by B C148680)
B3GNT7 is a glycosyltransferase which may play a role in elongation of the keratan sulfate carbohydrate backbone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta 1,3-N-acetylglucosaminyltransferase 7; beta-1,3-Gn-T7; beta-1,3-N-acetylglucosaminyltransferase 7; beta3Gn-T7; BGnT-7; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 7
Gene Aliases: B3GNT7; beta-3GnT7; beta3GnT7; C330001H22Rik
UniProt ID: (Human) Q8NFL0, (Mouse) Q8K0J2
Entrez Gene ID: (Human) 93010, (Mouse) 227327
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support