Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAFPCRRSLT AKTLACLLVG VSFLALQQWF LQAPRSPREE RSPQEETPEG PTDAPAADEP PSELVPGPPC VANASANATA DFEQLPARIQ DFLRYRHCRH FPLLWDAPAK CAGGRGVFLL LAVKSAPEHY ERRELIRRTW GQERSYGGRP VRRLFLLGTP GPEDEARAER LAELVALEAR EHGDVLQWAF ADTFLNLTLK HLHLLDWLAA RCPHARFLLS GDDDVFVHTA NVVRFLQAQP PGRHLFSGQL MEGSVPIRDS WSKYFVPPQL FPGSAYPVYC SGGGFLLSGP TARALRAAAR HTPLFPIDDA YMGMCLERAG LAPSGHEGIR PFGVQLPGAQ QSSFDPCMYR ELLLVHRFAP YEMLLMWKAL HSPALSCDRG HRVS
The protein encoded by this gene is a beta-1,3-N-acetylglucosaminyltransferase that adds an N-acetylglucosamine moiety to N-acetylgalactosamine-modified serine or threonine. The encoded enzyme is responsible for creating the core 3 structure of O-glycans, which are important components of mucin-type glycoproteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acetylgalactosaminyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase; beta-1,3-N-acetylglucosaminyltransferase 6; beta-1,3-N-acetylglucosaminyltransferase protein; BGnT-6; Core 3 synthase; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Gene Aliases: B3Gn-T6; B3GNT6; beta-1,3-Gn-T6; beta3Gn-T6; BGnT-6
UniProt ID: (Human) Q6ZMB0
Entrez Gene ID: (Human) 192134
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support