Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MWLGRRALCA LVLLLACASL GLLYASTRDA PGLRLPLAPW APPQSPRRPE LPDLAPEPRY AHIPVRIKEQ VVGLLAWNNC SCESSGGGLP LPFQKQVRAI DLTKAFDPAE LRAASATREQ EFQAFLSRSQ SPADQLLIAP ANSPLQYPLQ GVEVQPLRSI LVPGLSLQAA SGQEVYQVNL TASLGTWDVA GEVTGVTLTG EGQADLTLVS PGLDQLNRQL QLVTYSSRSY QTNTADTGAR PGWRDGQAGQ TEKNQKGWSG QMAEGMGGIW AMARAVQPHN GCFNWTSRAR GRKGAFVHLG LEQARGKPEP WVCLPFRPTV GGPRKRLV
GM2 and GD2 gangliosides are sialic acid-containing glycosphingolipids. GalNAc-T is the enzyme involved in the biosynthesis of G(M2) and G(D2) glycosphingolipids. GalNAc-T catalyzes the transfer of GalNAc into G(M3) and G(D3) by a beta-1,4 linkage, resulting in the synthesis of G(M2) and G(D2), respectively. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (N-acetylneuraminyl)-galactosylglucosylceramide; Beta-1,4 N-acetylgalactosaminyltransferase 1; beta-1,4-N-acetyl-galactosaminyl transferase 1; beta1,4GalNAc-T; GalNAc-T; GD2 synthase, GM2 synthase; GM2/GD2 synthase; UDP-Gal:betaGlcNAc beta-1,4-N-acetylgalactosaminyltransferase transferase 1; UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide N-acetylgalactosaminyltransferase (GalNAc-T)
Gene Aliases: B4GALNT1; GALGT; GalNAc-T; GALNACT; SIAT2; SPG26
UniProt ID: (Human) Q00973
Entrez Gene ID: (Human) 2583
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support